2-Chloro-5-(trifluoromethyl)pyrazine 97%

Supplier: Thermo Scientific Chemicals
Danger

H33863-MD H33863-03 H33863-MD H33863-03
AAH33863-03EA 186.99 USD
CAAAH33863-03 AAH33863-03 AAH33863-MD CAAAH33863-MD
2-Chloro-5-(trifluoromethyl)pyrazine 97%
2-Chloro-5-(trifluoromethyl)pyrazine
Formula: C₅H₂ClF₃N₂
Boiling Pt: 85 °C/112 mm
Storage Temperature: Ambient
MDL Number: MFCD13189478
CAS Number: 799557-87-2
UN: 2922
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR