3-Amino-4-hydroxybenzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Amino-4-cyanophenol
H33856-MD
H33856-06
H33856-06
H33856-MD
H33856-03
H33856-03
AAH33856-MDEA
72.02
USD
AAH33856-MD
AAH33856-06
CAAAH33856-03
CAAAH33856-MD
AAH33856-03
CAAAH33856-06
3-Amino-4-hydroxybenzonitrile 97%
3-Amino-4-hydroxybenzonitrile
Formula:
C₇H₆N₂O MW: 134.14 g/mol Melting Pt: 151…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234273 CAS Number: 14543-43-2 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...