2-Fluoro-4-(pentafluorothio)phenylacetic acid 97%

Warning

Synonyms: 2-Fluoro-4-(pentafluorosulfur)phenylacetic acid

H33834-MD H33834-03 H33834-03 H33834-MD
AAH33834-MDEA 95.46 USD
AAH33834-MD AAH33834-03 CAAAH33834-MD CAAAH33834-03
2-Fluoro-4-(pentafluorothio)phenylacetic acid 97%
2-Fluoro-4-(pentafluorothio)phenylacetic acid
Formula: C₈H₆F₆O₂S
MW: 280.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652509
CAS Number: 1240257-93-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR