4-Methoxy-3-(trifluoromethoxy)aniline 97%

Warning

H33826-06 H33826-03 H33826-06 H33826-03
AAH33826-03EA 109.44 USD
CAAAH33826-03 AAH33826-03 AAH33826-06 CAAAH33826-06
4-Methoxy-3-(trifluoromethoxy)aniline 97%
4-Methoxy-3-(trifluoromethoxy)aniline
Formula: C₈H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD18390012
CAS Number: 647855-21-8
UN: 2810
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR