4-Chloro-2-(trifluoromethoxy)benzyl bromide 97%
|
Danger
|
H33801-MD
H33801-03
H33801-MD
H33801-03
AAH33801-03EA
205.35
USD
CAAAH33801-03
CAAAH33801-MD
AAH33801-03
AAH33801-MD
4-Chloro-2-(trifluoromethoxy)benzyl bromide 97%
4-Chloro-2-(trifluoromethoxy)benzyl bromide
Formula:
C₈H₅BrClF₃O Storage Temperature: Ambient |
MDL Number:
MFCD18393833 CAS Number: 1261606-06-7 UN: 3265 ADR: "8",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...