5-Chloro-2-phenylimidazole-4-carboxaldehyde 97%

Warning

Synonyms: 5-Chloro-2-phenyl-1H-imidazole-4-carboxaldehyde

H33790-MD H33790-03 H33790-03 H33790-MD
AAH33790-MDEA 55.37 USD
CAAAH33790-03 AAH33790-MD CAAAH33790-MD AAH33790-03
5-Chloro-2-phenylimidazole-4-carboxaldehyde 97%
5-Chloro-2-phenylimidazole-4-carboxaldehyde
Formula: C₁₀H₇ClN₂O
MW: 206.629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01935953
CAS Number: 60367-52-4

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR