2-Methyl-5-(trifluoromethoxy)benzaldehyde 96%

Warning

H33783-03 H33783-06 H33783-06 H33783-03
AAH33783-03EA 79.35 USD
AAH33783-03 CAAAH33783-03 CAAAH33783-06 AAH33783-06
2-Methyl-5-(trifluoromethoxy)benzaldehyde 96%
2-Methyl-5-(trifluoromethoxy)benzaldehyde
Formula: C₉H₇F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD18399706
CAS Number: 195622-40-3

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR