Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-5-trifluoromethyl-o-phenylenediamine ≥95%

Supplier: Alfa Aesar
Danger

H33761-03 H33761-MD H33761-06 H33761-06 H33761-MD H33761-03
AAH33761-03EA 210.12 USD InStock
AAH33761-03 AAH33761-MD CAAAH33761-06 AAH33761-06 CAAAH33761-MD CAAAH33761-03
4-Chloro-5-trifluoromethyl-o-phenylenediamine ≥95%
4-Chloro-5-trifluoromethyl-o-phenylenediamine


Formula: C₇H₆ClF₃N₂
MW: 210.584 g/mol
Melting Pt: 61-63°C
Storage Temperature: Ambient
MDL Number: MFCD00119550
CAS Number: 157590-59-5
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty