4-Amino-N,N-dimethylbenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals

H33747-06 H33747-03 H33747-03 H33747-06
AAH33747-06EA 281.99 USD
AAH33747-06 CAAAH33747-06 CAAAH33747-03 AAH33747-03
4-Amino-N,N-dimethylbenzenesulfonamide 97%
4-Amino-N,N-dimethylbenzenesulphonamide
Formula: C₈H₁₂N₂O₂S
MW: 200.26 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00031428
CAS Number: 1709-59-7
EINECS: 216-970-5

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR