3-Fluoro-5-(trifluoromethoxy)benzoyl chloride 97%

Danger

Synonyms: 3-fluoro-5-(trifluoromethoxy)benzoyl chloride

H33742-03 H33742-03 H33742-MD H33742-MD
AAH33742-MDEA 54.46 USD
CAAAH33742-03 AAH33742-MD AAH33742-03 CAAAH33742-MD
3-Fluoro-5-(trifluoromethoxy)benzoyl chloride 97%
3-Fluoro-5-(trifluoromethoxy)benzoyl chloride
Formula: C₈H₃ClF₄O₂
MW: 242.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18783222
CAS Number: 1352999-78-0
UN: 3265
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR