2-Methyl-5-(pentafluorothio)benzaldehyde 97%

Warning

H33735-MD H33735-03 H33735-MD H33735-03
AAH33735-MDEA 94.39 USD
AAH33735-MD CAAAH33735-MD CAAAH33735-03 AAH33735-03
2-Methyl-5-(pentafluorothio)benzaldehyde 97%
2-Methyl-5-(pentafluorothio)benzaldehyde
Formula: C₈H₇F₅OS
MW: 246.19
Storage Temperature: Ambient
MDL Number: MFCD16652476
CAS Number: 1240257-13-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR