3-Methoxy-2-(trifluoromethyl)benzonitrile 97%
|
Warning
|
H33717-03
H33717-MD
H33717-03
H33717-MD
AAH33717-MDEA
41.22
USD
CAAAH33717-03
AAH33717-MD
CAAAH33717-MD
AAH33717-03
3-Methoxy-2-(trifluoromethyl)benzonitrile 97%
3-Methoxy-2-(trifluoromethyl)benzonitrile
Formula:
C₉H₆F₃NO Melting Pt: 103…105 °C Storage Temperature: Ambient |
MDL Number:
MFCD13185684 CAS Number: 1214385-02-0 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...