6-Chloro-2,3-difluoroaniline 97%

Warning

Synonyms: 6-Chloro-2,3-difluoroaniline

H33705-03 H33705-03 H33705-06 H33705-06
AAH33705-06EA 528.36 USD
CAAAH33705-03 AAH33705-06 AAH33705-03 CAAAH33705-06
6-Chloro-2,3-difluoroaniline 97%
6-Chloro-2,3-difluoroaniline
Formula: C₆H₄ClF₂N
MW: 163.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201032
CAS Number: 1373920-77-4
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR