4-Benzyl-1-boc-2-methylpiperazine 97%

Warning

H33694-MD H33694-03 H33694-03 H33694-MD
AAH33694-03EA 74.23 USD
AAH33694-03 CAAAH33694-MD AAH33694-MD CAAAH33694-03
4-Benzyl-1-boc-2-methylpiperazine 97%
4-Benzyl-1-boc-2-methylpiperazine
Formula: C₁₇H₂₆N₂O₂
MW: 290.407 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD03265484
CAS Number: 120737-77-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR