Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate 97%

Warning

Synonyms: Methyl 3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate

H33685-06 H33685-MD H33685-MD H33685-06
AAH33685-MDEA 23.86 USD
CAAAH33685-MD CAAAH33685-06 AAH33685-MD AAH33685-06
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate 97%
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate
Formula: C₁₀H₉NO₄
MW: 207.185 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD03036646
CAS Number: 202195-67-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR