1-Bromo-2-ethoxy-4-nitrobenzene 98%

Warning

Synonyms: 2-Bromo-5-nitrophenetole

H33669-06 H33669-14 H33669-14 H33669-06
AAH33669-14EA 301.92 USD
AAH33669-14 CAAAH33669-06 AAH33669-06 CAAAH33669-14
1-Bromo-2-ethoxy-4-nitrobenzene 98%
1-Bromo-2-ethoxy-4-nitrobenzene
Formula: C8H8BrNO3
MW: 246.06 g/mol
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03790890
CAS Number: 423165-33-7

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR