6-Chloro-2,3-difluorobenzoic acid 97%

Warning

Synonyms: 6-Chloro-2,3-difluorobenzoic acid

H33595-06 H33595-06 H33595-03 H33595-03
AAH33595-06EA 244.2 USD
AAH33595-06 AAH33595-03 CAAAH33595-03 CAAAH33595-06
6-Chloro-2,3-difluorobenzoic acid 97%
6-Chloro-2,3-difluorobenzoic acid
Formula: C₇H₃ClF₂O₂
MW: 192.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368094
CAS Number: 887584-84-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR