1-Benzyl-3-methylpiperazine 95%

Warning

H33591-03 H33591-06 H33591-03 H33591-06
AAH33591-03EA 91.24 USD
AAH33591-03 CAAAH33591-06 AAH33591-06 CAAAH33591-03
1-Benzyl-3-methylpiperazine 95%
1-Benzyl-3-methylpiperazine
Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Boiling Pt: 125…126 °C/2 mm
Storage Temperature: Ambient
MDL Number: MFCD01570796
CAS Number: 3138-90-7

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR