4-Methoxy-3-(trifluoromethoxy)benzyl alcohol 97%

Warning

H33588-03 H33588-03 H33588-MD H33588-MD
AAH33588-03EA 847.9 USD
CAAAH33588-03 AAH33588-03 AAH33588-MD CAAAH33588-MD
4-Methoxy-3-(trifluoromethoxy)benzyl alcohol 97%
4-Methoxy-3-(trifluoromethoxy)benzyl alcohol
Formula: C₉H₉F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD18398472
CAS Number: 853771-91-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR