4-Chloro-2-(trifluoromethoxy)benzaldehyde 97%

Warning

H33581-03 H33581-MD H33581-MD H33581-03
AAH33581-MDEA 54.6 USD
CAAAH33581-MD AAH33581-MD AAH33581-03 CAAAH33581-03
4-Chloro-2-(trifluoromethoxy)benzaldehyde 97%
4-Chloro-2-(trifluoromethoxy)benzaldehyde
Formula: C₈H₄ClF₃O₂
MW: 224.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393772
CAS Number: 1261442-48-1

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR