4-n-Butoxy-2,6-difluorophenylacetic acid 97%

Warning

Synonyms: 4-Butoxy-2,6-difluorophenylacetic acid

H33578-03 H33578-MD H33578-03 H33578-MD
AAH33578-03EA 236.43 USD
AAH33578-03 CAAAH33578-03 AAH33578-MD CAAAH33578-MD
4-n-Butoxy-2,6-difluorophenylacetic acid 97%
4-n-Butoxy-2,6-difluorophenylacetic acid
Formula: C₁₂H₁₄F₂O₃
MW: 244.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201023
CAS Number: 1373921-12-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR