3-Methoxy-4-(trifluoromethyl)benzoic acid 97%

Warning

H33574-06 H33574-03 H33574-03 H33574-06
AAH33574-03EA 107.44 USD
AAH33574-03 AAH33574-06 CAAAH33574-03 CAAAH33574-06
3-Methoxy-4-(trifluoromethyl)benzoic acid 97%
3-Methoxy-4-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Melting Pt: 209…211 °C
Storage Temperature: Ambient
MDL Number: MFCD13185682
CAS Number: 276861-63-3

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR