Methyl-6-methylpicolinate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 6-methylpyridine-2-carboxylate
H33571-MD
H33571-03
H33571-03
H33571-MD
AAH33571-MDEA
54.54
USD
AAH33571-MD
AAH33571-03
CAAAH33571-MD
CAAAH33571-03
Methyl-6-methylpicolinate 96%
Methyl 6-methylpicolinate
Formula:
C₈H₉NO₂ MW: 151.17 g/mol Boiling Pt: 242 °C Storage Temperature: Ambient |
MDL Number:
MFCD06203670 CAS Number: 13602-11-4 EINECS: 237-085-0 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...