10-Isocyanatodecyltrimethoxysilane 95%
|
Danger
|
H33555-03
H33555-03
H33555-MD
H33555-MD
AAH33555-MDEA
93.55
USD
CAAAH33555-MD
AAH33555-MD
AAH33555-03
CAAAH33555-03
10-Isocyanatodecyltrimethoxysilane 95%
10-Isocyanatodecyltrimethoxysilane
Formula:
C₁₄H₂₉NO₄Si Boiling Pt: 120…125 °C/0.1 mm Storage Temperature: Ambient |
MDL Number:
MFCD19105613 CAS Number: 1310372-82-7 UN: 2810 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...