4-(5-Bromo-2-pyridyloxy)benzenesulfonyl chloride 95%

Danger

Synonyms: 4-(5-Bromo-2-pyridyloxy)benzenesulfonyl chloride

H33550-MD H33550-03 H33550-03 H33550-MD
AAH33550-03EA 356.31 USD
CAAAH33550-MD AAH33550-03 CAAAH33550-03 AAH33550-MD
4-(5-Bromo-2-pyridyloxy)benzenesulfonyl chloride 95%
4-(5-Bromo-2-pyridyloxy)benzenesulfonyl chloride
Formula: C₁₁H₇BrClNO₃S
MW: 348.60 g/mol
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD21602384
CAS Number: 1312100-75-6
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR