2,4-Dibromo-5-(trifluoromethoxy)anisole 97%
|
Warning
|
H33545-06
H33545-06
H33545-03
H33545-03
AAH33545-06EA
192.03
USD
AAH33545-06
AAH33545-03
CAAAH33545-03
CAAAH33545-06
2,4-Dibromo-5-(trifluoromethoxy)anisole 97%
2,4-Dibromo-5-(trifluoromethoxy)anisole
Formula:
C₈H₅Br₂F₃O₂ MW: 349.93 Storage Temperature: Ambient |
MDL Number:
MFCD22201036 CAS Number: 1373920-68-3 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...