4-Methoxy-3-(trifluoromethoxy)benzoic acid 97%

Warning

H33542-06 H33542-06 H33542-03 H33542-03
AAH33542-03EA 123.21 USD
CAAAH33542-06 AAH33542-03 AAH33542-06 CAAAH33542-03
4-Methoxy-3-(trifluoromethoxy)benzoic acid 97%
4-Methoxy-3-(trifluoromethoxy)benzoic acid
Formula: C₉H₇F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD18398451
CAS Number: 647855-22-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR