4-Chloro-3-ethoxy-2-fluoroaniline 97%

Warning

H33541-03 H33541-MD H33541-03 H33541-MD
AAH33541-03EA 269.73 USD
AAH33541-03 CAAAH33541-03 CAAAH33541-MD AAH33541-MD
4-Chloro-3-ethoxy-2-fluoroaniline 97%
4-Chloro-3-ethoxy-2-fluoroaniline
Formula: C₈H₉ClFNO
Storage Temperature: Ambient
MDL Number: MFCD19687218
CAS Number: 1323966-31-9
UN: 2810
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR