3'-(Pentafluorothio)acetophenone 97%
|
Warning
|
H33532-03
H33532-06
H33532-03
H33532-06
AAH33532-06EA
396.27
USD
AAH33532-06
AAH33532-03
CAAAH33532-06
CAAAH33532-03
3'-(Pentafluorothio)acetophenone 97%
3'-(Pentafluorothio)acetophenone
Formula:
C₈H₇F₅OS Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD16652407 CAS Number: 159896-31-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...