4-Chloro-2-fluoro-3-methoxycinnamic acid 97%

Warning

Synonyms: 4-Chloro-2-fluoro-3-methoxycinnamic acid

H33530-MD H33530-MD H33530-03 H33530-03
AAH33530-03EA 138.75 USD
AAH33530-03 CAAAH33530-03 CAAAH33530-MD AAH33530-MD
4-Chloro-2-fluoro-3-methoxycinnamic acid 97%
4-Chloro-2-fluoro-3-methoxycinnamic acid
Formula: C₁₀H₈ClFO₃
MW: 230.62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687212
CAS Number: 1353001-74-7

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR