6'-Chloro-2',3'-difluoroacetophenone 97%
|
Warning
|
H33516-03
H33516-06
H33516-06
H33516-03
AAH33516-03EA
123.21
USD
CAAAH33516-06
AAH33516-03
CAAAH33516-03
AAH33516-06
6'-Chloro-2',3'-difluoroacetophenone 97%
6'-Chloro-2',3'-difluoroacetophenone
Formula:
C₈H₅ClF₂O Storage Temperature: Ambient |
MDL Number:
MFCD22201051 CAS Number: 1373920-86-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...