2-Fluoro-5-methyl-4-(trifluoromethyl)benzoic acid 97%
|
Warning
|
H33505-06
H33505-03
H33505-06
H33505-03
AAH33505-03EA
126.54
USD
CAAAH33505-06
AAH33505-03
CAAAH33505-03
AAH33505-06
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoic acid 97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoic acid
Formula:
C₉H₆F₄O₂ Storage Temperature: Ambient |
MDL Number:
MFCD19687178 CAS Number: 1323955-61-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...