4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide 97%

Danger

Synonyms: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide

H33496-06 H33496-06 H33496-03 H33496-03
AAH33496-06EA 528.36 USD
CAAAH33496-06 AAH33496-06 CAAAH33496-03 AAH33496-03
4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzyl bromide
Formula: C₉H₇BrF₄
MW: 271.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201072
CAS Number: 1373921-09-5
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR