(2-Fluoro-4-nitrophenyl)methanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Fluoro-4-nitrobenzyl alcohol

H33494-MD H33494-MD H33494-03 H33494-03
AAH33494-MDEA 38.18 USD
CAAAH33494-MD CAAAH33494-03 AAH33494-MD AAH33494-03
(2-Fluoro-4-nitrophenyl)methanol 97%
(2-Fluoro-4-nitrophenyl)methanol
Formula: C₇H₆FNO₃
MW: 171.127 g/mol
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD11110168
CAS Number: 660432-43-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR