4-Methoxy-3-(trifluoromethoxy)benzamide 97%

Warning

Synonyms: 4-methoxy-3-(trifluoromethoxy)benzamide

H33492-MD H33492-03 H33492-MD H33492-03
AAH33492-MDEA 34.46 USD
CAAAH33492-03 AAH33492-MD CAAAH33492-MD AAH33492-03
4-Methoxy-3-(trifluoromethoxy)benzamide 97%
4-Methoxy-3-(trifluoromethoxy)benzamide
Formula: C₉H₈F₃NO₃
MW: 235.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18398425
CAS Number: 1261646-41-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR