4-(4-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride 97%

Danger

Synonyms: 4-{[4-(Trifluoromethyl)pyridin-2-yl]oxy}benzenesulphonyl chloride

H33480-MD H33480-03 H33480-MD H33480-03
AAH33480-MDEA 67.73 USD
CAAAH33480-03 AAH33480-MD AAH33480-03 CAAAH33480-MD
4-(4-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride 97%
4-(4-Trifluoromethyl-2-pyridyloxy)benzenesulfonyl chloride
Formula: C₁₂H₇ClF₃NO₃S
MW: 337.71 g/mol
Melting Pt: 77…79 °C
Storage Temperature: Ambient
MDL Number: MFCD11109522
CAS Number: 1227955-11-4
UN: 3261
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR