2-Methyl-3-(trifluoromethyl)pyrazine 96%

Warning

Synonyms: 2-Methyl-3-trifluoromethylpyrazine

H33449-03 H33449-MD H33449-MD H33449-03
AAH33449-MDEA 143.19 USD
AAH33449-MD CAAAH33449-03 CAAAH33449-MD AAH33449-03
2-Methyl-3-(trifluoromethyl)pyrazine 96%
2-Methyl-3-(trifluoromethyl)pyrazine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Boiling Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD14525502
CAS Number: 1204295-56-6

Order Now

Specification Test Results

Assay Minimum 96

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR