3-Bromo-4-fluoro-5-iodobenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H33441-03
H33441-03
H33441-MD
H33441-MD
AAH33441-MDEA
126.16
USD
AAH33441-MD
AAH33441-03
CAAAH33441-MD
CAAAH33441-03
3-Bromo-4-fluoro-5-iodobenzaldehyde 97%
3-Bromo-4-fluoro-5-iodobenzaldehyde
Formula:
C₇H₃BrFIO Storage Temperature: Ambient |
MDL Number:
MFCD20486165 CAS Number: 1356113-37-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...