Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Alfa Aesar
Warning

H33404-03 H33404-MD H33404-MD H33404-03
AAH33404-03EA 148.32 USD InStock
AAH33404-03 AAH33404-MD CAAAH33404-MD CAAAH33404-03
1-Bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene ≥95%
1-Bromo-4-fluoro-2-(2,2,2-trifluoroethyl)benzene


Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Boiling Pt: 76°/15mm
Storage Temperature: Ambient
MDL Number: MFCD11226691
CAS Number: 1186194-80-8
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty