2-Methyl-5-(trifluoromethoxy)phenylacetonitrile 97%

Warning

Synonyms: 2-Methyl-5-(trifluoromethoxy)phenylacetonitrile

H33402-03 H33402-MD H33402-MD H33402-03
AAH33402-MDEA 46.2 USD
AAH33402-MD CAAAH33402-MD AAH33402-03 CAAAH33402-03
2-Methyl-5-(trifluoromethoxy)phenylacetonitrile 97%
2-Methyl-5-(trifluoromethoxy)phenylacetonitrile
Formula: C₁₀H₈F₃NO
MW: 215.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399826
CAS Number: 1261683-78-6
UN: 3276
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR