4-Chloro-2-fluoro-3-methoxybenzamide 97%

Warning

Synonyms: 4-chloro-2-fluoro-3-methoxybenzamide

H33392-03 H33392-MD H33392-MD H33392-03
AAH33392-MDEA 67.49 USD
AAH33392-MD CAAAH33392-MD CAAAH33392-03 AAH33392-03
4-Chloro-2-fluoro-3-methoxybenzamide 97%
4-Chloro-2-fluoro-3-methoxybenzamide
Formula: C₈H₇ClFNO₂
MW: 203.60 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687206
CAS Number: 1323966-12-6

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR