2-Fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride 97%

Danger

Synonyms: 2-fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride

H33390-03 H33390-MD H33390-MD H33390-03
AAH33390-MDEA 54.6 USD
AAH33390-MD CAAAH33390-MD CAAAH33390-03 AAH33390-03
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride 97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoyl chloride
Formula: C₉H₅ClF₄O
MW: 240.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19687193
CAS Number: 1323966-18-2
UN: 3265
ADR: "8",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR