2-Chloro-4-(trifluoromethyl)benzyl alcohol 97%

Warning

H33376-MD H33376-03 H33376-03 H33376-MD
AAH33376-03EA 316.35 USD
CAAAH33376-MD AAH33376-03 CAAAH33376-03 AAH33376-MD
2-Chloro-4-(trifluoromethyl)benzyl alcohol 97%
2-Chloro-4-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Melting Pt: 50…52 °C
Storage Temperature: Ambient
MDL Number: MFCD04972757
CAS Number: 56456-51-0

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR