2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile 97%
|
Warning
|
H33354-06
H33354-03
H33354-03
H33354-06
AAH33354-06EA
528.36
USD
AAH33354-06
CAAAH33354-03
CAAAH33354-06
AAH33354-03
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzonitrile
Formula:
C₉H₅F₄NO Storage Temperature: Ambient |
MDL Number:
MFCD22201044 CAS Number: 1373920-99-0 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...