3-Fluoro-5-(pentafluorosulfur)benzonitrile 97%

Warning

Synonyms: 3-Fluoro-5-(pentafluorothio)benzonitrile

H33352-03 H33352-MD H33352-MD H33352-03
AAH33352-03EA 275.28 USD
CAAAH33352-MD AAH33352-03 CAAAH33352-03 AAH33352-MD
3-Fluoro-5-(pentafluorosulfur)benzonitrile 97%
3-Fluoro-5-(pentafluorosulfur)benzonitrile
Formula: C₇H₃ClF₅NS
MW: 247.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201029
CAS Number: 159727-28-3
UN: 3439
ADR: "6.1",III

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR