4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde 97%

Warning

Synonyms: 4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde

H33320-06 H33320-03 H33320-03 H33320-06
AAH33320-06EA 190.92 USD
AAH33320-06 CAAAH33320-06 AAH33320-03 CAAAH33320-03
4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde 97%
4-Fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde
Formula: C₉H₆F₄O
MW: 206.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22201063
CAS Number: 1373921-14-2

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR