4-Methoxy-3-(trifluoromethoxy)benzonitrile 97%
|
Warning
|
H33310-MD
H33310-MD
H33310-03
H33310-03
AAH33310-03EA
158.73
USD
AAH33310-03
CAAAH33310-MD
AAH33310-MD
CAAAH33310-03
4-Methoxy-3-(trifluoromethoxy)benzonitrile 97%
4-Methoxy-3-(trifluoromethoxy)benzonitrile
Formula:
C₉H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD18398458 CAS Number: 1261653-03-5 UN: 3439 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...