Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3'-Fluoro-5'-(pentafluorothio)acetophenone 97%

Supplier: Alfa Aesar
Warning

H33302-03 H33302-MD H33302-MD H33302-03
AAH33302-03EA 267.8 USD InStock
AAH33302-03 CAAAH33302-MD AAH33302-MD CAAAH33302-03
3'-Fluoro-5'-(pentafluorothio)acetophenone 97%
3'-Fluoro-5'-(pentafluorothio)acetophenone


Formula: C₈H₆F₆OS
MW: 264,18
Storage Temperature: Ambient
MDL Number: MFCD16652464
CAS Number: 1240257-57-1
EINECS: 000-000-0
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty