2'-Fluorobiphenyl-4-carboxylic acid 97%

Warning

Synonyms: 2'-Fluoro-[1,1'-biphenyl]-4-carboxylic acid

H33301-03 H33301-06 H33301-03 H33301-06
AAH33301-06EA 177.6 USD
CAAAH33301-06 AAH33301-06 AAH33301-03 CAAAH33301-03
2'-Fluorobiphenyl-4-carboxylic acid 97%
2'-Fluorobiphenyl-4-carboxylic acid
Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
Melting Pt: 230…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00278732
CAS Number: 365-12-8

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR