4-Chloro-2-fluoro-3-methoxybenzaldehyde 97%

Warning

H33274-06 H33274-03 H33274-06 H33274-03
AAH33274-03EA 151.17 USD
AAH33274-03 CAAAH33274-06 CAAAH33274-03 AAH33274-06
4-Chloro-2-fluoro-3-methoxybenzaldehyde 97%
4-Chloro-2-fluoro-3-methoxybenzaldehyde
Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD19687172
CAS Number: 1002344-97-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR